Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rffl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004068.1 | NP_001004068.1 | 6 | 1488 | ||
XM_006246969.2 | XP_006247031.1 | 7 | 1646 | |||
XM_006246970.2 | XP_006247032.1 | 7 | 1643 | |||
XM_006246973.2 | XP_006247035.1 | 7 | 1883 | |||
XM_006246974.2 | XP_006247036.1 | 7 | 1473 | |||
XM_006246975.2 | XP_006247037.2 | 8 | 2454 | |||
XM_006246977.2 | XP_006247039.1 | 7 | 2362 | |||
XM_006246978.2 | XP_006247040.1 | 7 | 1880 | |||
XM_006246979.2 | XP_006247041.1 | 7 | 1469 | |||
XM_008767954.1 | XP_008766176.1 | 8 | 2451 | |||
XM_008767955.1 | XP_008766177.1 | 8 | 2636 | |||
XM_008767956.1 | XP_008766178.1 | 7 | 2371 | |||
GenBank mRNA | BC079216.1 | - | 6 | 1488 | 7131890 | |
FQ210900.1 | - | 7 | 1354 |