Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004072.2 | NP_001004072.2 | 11 | 2533 | ||
GenBank mRNA | BC079369.1 | - | 11 | 2525 | ||
BC098897.1 | - | 11 | 2533 | 7103543 | ||
FQ214890.1 | - | 1 | 863 | |||
FQ235262.1 | - | 1 | 790 | |||
Z12158.1 | - | 11 | 2552 |