Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004077.1 | NP_001004077.1 | 1 | 416 | ||
GenBank mRNA | BC079449.1 | - | 1 | 416 | 7115796 |