Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004078.1 | NP_001004078.1 | 11 | 1536 | ||
GenBank mRNA | BC079441.1 | - | 11 | 1536 | 7115414 | |
FQ229253.1 | - | 6 | 755 |