Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pon3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004086.1 | NP_001004086.1 | 9 | 1270 | ||
GenBank mRNA | BC079466.1 | - | 9 | 1270 | 7122531 | |
FQ218322.1 | - | 6 | 1013 | |||
FQ218895.1 | - | 6 | 1010 | |||
FQ219524.1 | - | 6 | 1010 |