Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il17re | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004091.1 | NP_001004091.1 | 4 | 407 | ||
GenBank mRNA | BC078742.1 | - | 4 | 407 | 7123764 |