Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207033.1 | NP_001193962.1 | 16 | 3174 | ||
NM_001207034.1 | NP_001193963.1 | 14 | 2874 | |||
NM_001286572.2 | NP_001273501.1 | 16 | 2535 | |||
NM_014771.3 | NP_055586.1 | 16 | 3174 | |||
XM_011545997.1 | XP_011544299.1 | 16 | 2515 | |||
GenBank mRNA | AB014561.1 | - | 16 | 2446 | ||
AF122819.1 | - | 16 | 2450 | |||
AK027406.1 | - | 14 | 2164 | |||
AK291666.1 | - | 16 | 2464 | |||
AK293710.1 | - | 6 | 677 | |||
AK294258.1 | - | 6 | 759 | |||
AK297531.1 | - | 10 | 1580 | |||
AK304521.1 | - | 3 | 181 | |||
BC004527.2 | - | 2 | 157 | |||
BC006133.1 | - | 16 | 2441 | 3628183 | ||
BC011769.2 | - | 16 | 2441 | 3607432 | ||
BC018647.2 | - | 16 | 2441 | 2967275 | ||
BC030802.1 | - | 8 | 1335 | |||
BC113827.1 | - | 3 | 360 | |||
BX640763.1 | - | 13 | 1808 | |||
CR627431.1 | - | 16 | 2503 | |||
KU178492.1 | - | 15 | 2354 | |||
KU178493.1 | - | 15 | 2354 |