Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004140.2 | NP_001004140.2 | 4 | 542 | ||
GenBank mRNA | AK034534.1 | - | 4 | 497 | ||
AK132463.1 | - | 4 | 530 | |||
AY692438.1 | - | 4 | 542 | |||
BC145666.1 | - | 4 | 535 | 40130969 | ||
BC146023.1 | - | 4 | 535 | 40131326 |