Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004143.4 | NP_001004143.2 | 13 | 2442 | ||
GenBank mRNA | AK034492.1 | - | 13 | 2417 | ||
AK084022.1 | - | 9 | 1601 | |||
AK155071.1 | - | 13 | 2443 | |||
AK160992.1 | - | 13 | 2351 | |||
AK191821.1 | - | 1 | 77 | |||
AK193842.1 | - | 1 | 77 | |||
AK198335.1 | - | 1 | 77 | |||
AK199010.1 | - | 1 | 77 | |||
AK220368.1 | - | 13 | 2403 | |||
BC026701.1 | - | 6 | 1214 | |||
BC052540.1 | - | 9 | 1736 | |||
BC058419.1 | - | 6 | 1261 | |||
BC080737.1 | - | 13 | 2313 | 30431735 |