Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AA467197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004174.1 | NP_001004174.1 | 5 | 493 | ||
GenBank mRNA | AK141297.1 | - | 4 | 596 | ||
AK150561.1 | - | 5 | 483 | |||
BC049633.1 | - | 5 | 493 | 6774154 |