Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem178b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004182.3 | NP_001004182.1 | 1, 2 | 129 | ||
GenBank mRNA | AK084030.1 | - | 1, 2 | 130 |