Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004200.1 | NP_001004200.1 | 3 | 292 | ||
GenBank mRNA | BC079143.1 | - | 3 | 292 | 7128107 | |
FQ228882.1 | - | 3 | 333 |