Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004216.1 | NP_001004216.1 | 11, 12 | 1463 | ||
GenBank mRNA | AY573239.1 | - | 11, 12 | 1463 |