Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fuca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004218.1 | NP_001004218.1 | 1 | 276 | ||
GenBank mRNA | BC078933.1 | - | 1 | 276 | 7107565 |