Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam98c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198584.1 | NP_001185513.1 | 4 | 448 | ||
XM_006228717.2 | XP_006228779.1 | 4 | 478 | |||
GenBank mRNA | BC079080.1 | - | 3 | 486 | 7122702 | |
CO564588.1 | - | 2 | 225 | |||
CO570848.1 | - | 4 | 448 | |||
DV214726.1 | - | 4 | 473 |