Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004249.1 | NP_001004249.1 | 3 | 732 | ||
GenBank mRNA | BC079220.1 | - | 3 | 732 | 7131966 | |
FQ223087.1 | - | 3 | 670 |