Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004260.1 | NP_001004260.1 | 11 | 1425 | ||
XM_006230775.2 | XP_006230837.1 | 11 | 1484 | |||
GenBank mRNA | BC079159.1 | - | 11 | 1425 | 7129231 | |
FQ228759.1 | - | 11 | 1454 |