Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pyroxd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004261.1 | NP_001004261.1 | 7, 8 | 698 | ||
XM_006231394.2 | XP_006231456.1 | 7, 8 | 969 | |||
XM_006231396.2 | XP_006231458.1 | 7, 8 | 970 | |||
XM_008760424.1 | XP_008758646.1 | 7, 8 | 972 | |||
XM_008760425.1 | XP_008758647.1 | 7, 8 | 735 | |||
GenBank mRNA | BC079368.1 | - | 7, 8 | 698 | 7109628 |