Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCHSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014824.2 | NP_055639.2 | 4, 5 | 471 | ||
XM_011545409.1 | XP_011543711.1 | 4, 5 | 353 | |||
XM_011545410.2 | XP_011543712.1 | 4, 5 | 286 | |||
XM_017018632.1 | XP_016874121.1 | 4, 5 | 530 | |||
GenBank mRNA | AB018312.1 | - | 3, 4 | 321 | ||
AK297928.1 | - | 4, 5 | 355 | |||
BC010394.1 | - | 4, 5 | 218 | 4091973 | ||
CD358961.1 | - | 4, 5 | 485 |