Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004268.1 | NP_001004268.1 | 2, 3 | 1108 | ||
XM_006239015.2 | XP_006239077.1 | 2, 3 | 2149 | |||
XM_006239016.1 | XP_006239078.1 | 2, 3 | 1333 | |||
XM_006239017.2 | XP_006239079.1 | 2, 3 | 1946 | |||
XM_006239018.2 | XP_006239080.1 | 2, 3 | 1239 | |||
XM_008764133.1 | XP_008762355.1 | 1, 2 | 1575 | |||
GenBank mRNA | BC079148.1 | - | 2, 3 | 1108 | 7128510 |