Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snupn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004270.1 | NP_001004270.1 | 8 | 945 | ||
XM_006243124.1 | XP_006243186.1 | 9 | 1571 | |||
XM_006243125.1 | XP_006243187.1 | 9 | 1232 | |||
XM_006243126.1 | XP_006243188.1 | 9 | 1251 | |||
XM_006243127.2 | XP_006243189.1 | 9 | 1195 | |||
GenBank mRNA | BC079451.1 | - | 8 | 945 | 7121061 |