Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snupn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004270.1 | NP_001004270.1 | 5 | 542 | ||
XM_006243124.1 | XP_006243186.1 | 6 | 1168 | |||
XM_006243125.1 | XP_006243187.1 | 6 | 829 | |||
XM_006243126.1 | XP_006243188.1 | 6 | 848 | |||
XM_006243127.2 | XP_006243189.1 | 6 | 792 | |||
XM_006243128.2 | XP_006243190.1 | 6 | 1168 | |||
GenBank mRNA | BC079451.1 | - | 5 | 542 | 7121061 |