Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004277.2 | NP_001004277.1 | 5 | 675 | ||
GenBank mRNA | AY490816.1 | - | 5 | 647 | ||
BC098894.1 | - | 5 | 675 | 7099635 |