Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014830.2 | NP_055645.1 | 2 | 364 | ||
GenBank mRNA | AB002350.1 | - | 2 | 297 | ||
AK292468.1 | - | 2 | 364 | |||
BC140890.1 | - | 1 | 267 | 9021758 | ||
BC146867.1 | - | 1 | 267 | 9056795 | ||
BC152461.1 | - | 2 | 297 | 8862597 |