Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR1CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027484.2 | 4 | 596 | |||
GenBank mRNA | EB386870.1 | - | 2, 3 | 329 |
Gene Symbol: FCGR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004340.3 | NP_001004340.2 | 2 | 320 | ||
NM_001017986.3 | NP_001017986.1 | 4 | 596 | |||
NM_001244910.1 | NP_001231839.1 | 4 | 596 | |||
NR_045213.1 | 4 | 596 | ||||
XM_017000661.1 | XP_016856150.1 | 4 | 599 | |||
XM_017000662.1 | XP_016856151.1 | 3 | 341 | |||
GenBank mRNA | AK307244.1 | - | 4 | 596 | ||
AK308886.1 | - | 4 | 582 | |||
CD677904.1 | - | 1 | 134 | |||
L03419.1 | - | 4 | 546 | |||
L03420.1 | - | 2 | 270 |