Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TECPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172631.2 | NP_001166102.1 | 2 | 372 | ||
NM_014844.4 | NP_055659.2 | 2 | 372 | |||
GenBank mRNA | AB002327.1 | - | 2 | 321 | ||
AK127721.1 | - | 2 | 805 | |||
AK296395.1 | - | 2 | 223 | |||
BC136647.1 | - | 1 | 169 | 9020637 | ||
BC142667.1 | - | 2 | 205 | 40148157 | ||
BC142715.1 | - | 2 | 205 | 40148843 | ||
BC151230.1 | - | 2 | 345 | 8860116 | ||
BP365224.1 | - | 2 | 374 |