Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpgs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004361.2 | NP_001004361.2 | 7 | 1240 | ||
NM_001142697.1 | NP_001136169.1 | 7 | 986 | |||
GenBank mRNA | AK017728.1 | - | 7 | 992 | ||
AK171952.1 | - | 1 | 406 | |||
AK212422.1 | - | 1 | 32 | |||
BC060253.1 | - | 5 | 792 | |||
BC080777.1 | - | 7 | 996 | 30439612 |