Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004364.2 | NP_001004364.2 | 21 | 2405 | ||
NM_001098168.1 | NP_001091637.1 | 22 | 2414 | |||
NM_001135192.1 | NP_001128664.1 | 22 | 2405 | |||
GenBank mRNA | AK147622.1 | - | 22 | 2413 | ||
AK147656.1 | - | 22 | 2360 | |||
AK162242.1 | - | 1704 | ||||
AK172944.1 | - | 22 | 2226 | |||
BC080847.1 | - | 17 | 1967 | 6852788 | ||
BC096022.1 | - | 22 | 2182 | 30542525 |