Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxxc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004367.4 | NP_001004367.2 | 4 | 3263 | ||
GenBank mRNA | AK047627.1 | - | 1 | 453 | ||
BC065149.1 | - | 5 | 2949 | |||
BC082332.1 | - | 3 | 2480 | 30537505 |