Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tacc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004415.2 | NP_001004415.1 | 6782 | |||
NM_001004418.2 | NP_001004418.1 | 1667 | ||||
XM_006230398.2 | XP_006230460.1 | 3 | 1763 | |||
XM_008759883.1 | XP_008758105.1 | 3 | 1763 | |||
XM_008759884.1 | XP_008758106.1 | 7645 | ||||
GenBank mRNA | FQ223932.1 | - | 1 | 56 | ||
KJ143743.1 | - | 3 | 1133 |