Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tacc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004424.1 | NP_001004424.1 | 7 | 1052 | ||
XM_006251329.2 | XP_006251391.1 | 8 | 1405 | |||
XM_006251330.2 | XP_006251392.1 | 8 | 1408 | |||
XM_006251331.2 | XP_006251393.1 | 7 | 1067 |