Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SV2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323031.1 | NP_001309960.1 | 2 | 614 | ||
NM_001323032.1 | NP_001309961.1 | 2 | 923 | |||
NM_001323033.1 | NP_001309962.1 | 2 | 923 | |||
NM_001323034.1 | NP_001309963.1 | 2 | 923 | |||
NM_001323037.1 | NP_001309966.1 | 3 | 830 | |||
NM_001323038.1 | NP_001309967.1 | 2 | 742 | |||
NM_001323039.1 | NP_001309968.1 | 2 | 1015 | |||
NM_014848.5 | NP_055663.1 | 3 | 1011 | |||
XM_005254998.3 | XP_005255055.1 | 2 | 676 | |||
XM_017022761.1 | XP_016878250.1 | 2 | 627 | |||
XM_017022762.1 | XP_016878251.1 | 2 | 762 | |||
GenBank mRNA | AB018278.1 | - | 1 | 284 | ||
AF052188.1 | - | 2 | 538 | |||
BC030011.2 | - | 3 | 648 | 4826206 | ||
CR457086.1 | - | 1 | 120 |