Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METRNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004431.2 | NP_001004431.1 | 4 | 822 | ||
XM_017024523.1 | XP_016880012.1 | 4 | 1171 | |||
GenBank mRNA | AK093748.1 | - | 3 | 1393 | ||
BC050568.1 | - | 3 | 552 | |||
BC082252.1 | - | 4 | 822 | 6301543 | ||
BC118558.1 | - | 4 | 309 | |||
BC118638.1 | - | 4 | 309 | |||
DR760693.1 | - | 3 | 542 |