Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wapal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004436.4 | NP_001004436.2 | 2 | 576 | ||
NM_001301330.1 | NP_001288259.1 | 2 | 576 | |||
GenBank mRNA | AB167349.1 | - | 2 | 382 | ||
AK041191.1 | - | 2 | 310 | |||
AK046038.1 | - | 3 | 485 | |||
AK129105.1 | - | 2 | 576 | |||
BC139434.1 | - | 2 | 331 | 8862456 | ||
BC145625.1 | - | 2 | 331 | 9054193 | ||
BY732412.1 | - | 2 | 578 | |||
CJ134128.1 | - | 2 | 579 |