Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wapal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004436.4 | NP_001004436.2 | 3 | 1405 | ||
NM_001301330.1 | NP_001288259.1 | 3 | 1405 | |||
GenBank mRNA | AB167349.1 | - | 3 | 1211 | ||
AK041191.1 | - | 3 | 1139 | |||
AK045727.1 | - | 1 | 1723 | |||
AK129105.1 | - | 3 | 1405 | |||
BC038072.1 | - | 2 | 531 | |||
BC139434.1 | - | 3 | 1160 | 8862456 | ||
BC145625.1 | - | 3 | 1160 | 9054193 |