Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004446.1 | NP_001004446.1 | 4 | 702 | ||
XM_006231227.2 | XP_006231289.1 | 4 | 720 | |||
XM_008760331.1 | XP_008758553.1 | 4 | 980 | |||
XM_008760332.1 | XP_008758554.1 | 5 | 1028 | |||
XM_008760333.1 | XP_008758555.1 | 5 | 768 | |||
XM_008760334.1 | XP_008758556.1 | 4 | 800 | |||
XM_008760335.1 | XP_008758557.1 | 4 | 736 | |||
XM_008760336.1 | XP_008758558.1 | 4 | 1048 | |||
XM_008760337.1 | XP_008758559.1 | 3 | 598 | |||
XM_008760338.1 | XP_008758560.1 | 2 | 234 | |||
GenBank mRNA | AY643746.1 | - | 4 | 702 |