Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004447.1 | NP_001004447.1 | 14 | 2171 | ||
XM_008764277.1 | XP_008762499.1 | 14 | 2320 | |||
GenBank mRNA | AB168097.1 | - | 14 | 2171 | ||
BC089089.1 | - | 13 | 2090 | 7112067 |