Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEURL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005408.1 | NP_001005408.1 | 12 | 2193 | ||
NM_032442.2 | NP_115818.2 | 12 | 2193 | |||
GenBank mRNA | AB058690.1 | - | 12 | 2193 | ||
AL136870.1 | - | 12 | 2049 | |||
AL833570.1 | - | 12 | 1439 |