Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amica1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005421.4 | NP_001005421.3 | 6 | 745 | ||
GenBank mRNA | AA798428.1 | - | 264 | |||
AY093688.1 | - | 6 | 745 | |||
BC050133.1 | - | 4 | 632 | 30023491 |