Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqsec2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005475.2 | NP_001005475.1 | 5 | 1710 | ||
NM_001114664.2 | NP_001108136.1 | 5 | 2474 | |||
GenBank mRNA | AB275281.1 | - | 5 | 2474 | ||
AY451401.1 | - | 5 | 2467 |