Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127660.1 | NP_001121132.1 | 3 | 554 | ||
NM_014874.3 | NP_055689.1 | 4 | 699 | |||
XM_005263543.3 | XP_005263600.1 | 5 | 712 | |||
XM_005263545.3 | XP_005263602.1 | 4 | 562 | |||
XM_005263547.3 | XP_005263604.1 | 4 | 594 | |||
XM_005263548.3 | XP_005263605.1 | 5 | 729 | |||
GenBank mRNA | AF036536.2 | - | 4 | 568 | ||
AK289828.1 | - | 3 | 499 | |||
AY028429.1 | - | 3 | 272 | |||
BC017061.1 | - | 4 | 429 | 3901235 | ||
BU178912.1 | - | 4 | 494 | |||
D86987.1 | - | 4 | 568 |