Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmtk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005511.3 | NP_001005511.2 | 6 | 532 | ||
NM_001290990.1 | NP_001277919.1 | 5 | 687 | |||
GenBank mRNA | AB288873.1 | - | 6 | 526 | ||
BC057032.1 | - | 5 | 420 | |||
BC059845.1 | - | 6 | 741 | |||
BC083185.1 | - | 5 | 687 | 6855048 | ||
BC094377.1 | - | 6 | 532 | 30536822 |