Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005529.1 | NP_001005529.1 | 4, 5 | 594 | ||
XM_008767639.1 | XP_008765861.1 | 4, 5 | 645 | |||
GenBank mRNA | BC082093.1 | - | 4, 5 | 594 | 7134129 | |
FQ213119.1 | - | 4, 5 | 611 |