Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005535.2 | NP_001005535.1 | 2 | 414 | ||
NM_001190820.1 | NP_001177749.1 | 2 | 414 | |||
GenBank mRNA | BC081871.1 | - | 2 | 368 | 7106377 | |
DY310785.1 | - | 2 | 438 | |||
DY317721.1 | - | 2 | 414 |