Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srfbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005536.1 | NP_001005536.1 | 1 | 114 | ||
GenBank mRNA | BC082077.1 | - | 1 | 114 | 7133229 |