Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005541.1 | NP_001005541.1 | 8 | 789 | ||
GenBank mRNA | BC081963.1 | - | 8 | 789 | 7124985 | |
FQ212033.1 | - | 7 | 881 |