Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COBLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278458.1 | NP_001265387.1 | 5 | 657 | ||
NM_001278460.1 | NP_001265389.1 | 4 | 636 | |||
NM_001278461.1 | NP_001265390.1 | 3 | 596 | |||
NM_014900.4 | NP_055715.3 | 3 | 596 | |||
GenBank mRNA | AB023194.1 | - | 3 | 590 | ||
AK002054.1 | - | 4 | 538 | |||
AK225849.1 | - | 4 | 493 | |||
AK294937.1 | - | 5 | 657 | |||
BC071588.1 | - | 3 | 598 | 30345650 |