Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005902.1 | NP_001005902.1 | 7 | 689 | ||
GenBank mRNA | BC083579.1 | - | 7 | 689 | 7128126 |