Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005906.1 | NP_001005906.1 | 4 | 1537 | ||
XM_006245228.1 | XP_006245290.1 | 4 | 1166 | |||
GenBank mRNA | BC083617.1 | - | 4 | 1537 | 7133858 |