Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efemp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005907.2 | NP_001005907.1 | 10 | 1245 | ||
NM_001277341.1 | NP_001264270.1 | 11 | 1368 | |||
NM_001277346.1 | NP_001264275.1 | 9 | 1038 | |||
XM_006230742.2 | XP_006230804.1 | 11 | 1708 | |||
XM_006230743.1 | XP_006230805.1 | 11 | 1359 | |||
XM_006230744.2 | XP_006230806.1 | 11 | 1365 | |||
GenBank mRNA | BC083804.1 | - | 10 | 1183 | 7109182 |